accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOHOH

CAS RN 23470-00-0

IUPAC Name: 1,3-dihydroxypropan-2-yl hexadecanoate
Synonyms: 1,3-dihydroxypropan-2-yl hexadecanoate
Molecular Weight (g/mol): 330.51
Molecular Formula: C19H38O4
InChi Key: BBNYCLAREVXOSG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC(CO)CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results