accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 234108-73-7

H3CH3CCH3OONHNCl

CAS RN 234108-73-7

IUPAC Name: tert-butyl N-(2-chloropyridin-4-yl)carbamate
Synonyms: tert-butyl N-(2-chloropyridin-4-yl)carbamate
Molecular Weight (g/mol): 228.68
Molecular Formula: C10H13ClN2O2
InChi Key: SJCKHLJWAXGPGT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results