accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 23359-08-2

HOOO(E)

CAS RN 23359-08-2

IUPAC Name: (2E)-3-(4-formylphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(4-formylphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: LBOUHDMYVURTMA-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C1=CC=C(C=O)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)