accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOPOOCH3OCH3

CAS RN 2327-69-7

IUPAC Name: ethyl 4-(diethoxyphosphoryl)butanoate
Synonyms: ethyl 4-(diethoxyphosphoryl)butanoate
Molecular Weight (g/mol): 252.25
Molecular Formula: C10H21O5P
InChi Key: FYESTJSRASJZOY-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCP(=O)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results