accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 23165-53-9

CH3ClNCS

CAS RN 23165-53-9

IUPAC Name: 4-chloro-1-isothiocyanato-2-methylbenzene
Synonyms: 4-chloro-1-isothiocyanato-2-methylbenzene
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: XTYLRVPBHHRTMS-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)