CAS RN 23147-59-3
IUPAC Name:
3,6-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
Synonyms:
3,6-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
Molecular Weight (g/mol):
180.16
Molecular Formula:
C6H12O6
InChi Key:
HQAMJDIBKAKYMU-UHFFFAOYNA-N
SMILES:
OCC1OC(O)C(CO)OC1O
1
–
1
of
1
results
1
DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals
CAS: 23147-59-3 Molecular Formula: C6H12O6 Molecular Weight (g/mol): 180.16 InChI Key: HQAMJDIBKAKYMU-UHFFFAOYSA-N Synonym: 3,6-bis hydroxymethyl-1,4-dioxane-2,5-diol,pubchem10426,dl-glyceraldehyde dimer+,2,5-dihydroxy-3,6-dihydroxymethyl-1,4-dioxane PubChem CID: 251615 IUPAC Name: 3,6-bis(hydroxymethyl)-1,4-dioxane-2,5-diol SMILES: C(C1C(OC(C(O1)O)CO)O)O