accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 23135-50-4

OOHNOOH

CAS RN 23135-50-4

IUPAC Name: 5-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: QYYPKLYDFCYGPG-UHFFFAOYSA-N
SMILES: OC(=O)CCCCNC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)