accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 23132-21-0

CH3NBrNOO

CAS RN 23132-21-0

IUPAC Name: 2-bromo-3-methyl-5-nitropyridine
Synonyms: 2-bromo-3-methyl-5-nitropyridine
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: FZIQHPKXSLHGBZ-UHFFFAOYSA-N
SMILES: CC1=CC(=CN=C1Br)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results