accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2FFFF

CAS RN 231291-14-8

IUPAC Name: 1-[5-fluoro-2-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[5-fluoro-2-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 193.15
Molecular Formula: C8H7F4N
InChi Key: ZRDWPCJUMDGSCG-UHFFFAOYSA-N
SMILES: NCC1=CC(F)=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results