CAS RN 23112-96-1
IUPAC Name:
(2,6-dimethoxyphenyl)boronic acid
Synonyms:
2,6-dimethoxyphenylboronic acid
Molecular Weight (g/mol):
181.98
Molecular Formula:
C8H11BO4
InChi Key:
BKWVXPCYDRURMK-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(OC)=C1B(O)O
1
–
1
of
1
results
1
2,6-Dimethoxybenzeneboronic acid, 97+%
CAS: 23112-96-1 Molecular Formula: C8H11BO4 Molecular Weight (g/mol): 181.982 MDL Number: MFCD01318987 InChI Key: BKWVXPCYDRURMK-UHFFFAOYSA-N Synonym: 2,6-dimethoxyphenyl boronic acid,2,6-dimethoxyphenylboronicacid,2,6-dimethoxybenzeneboronic acid,2,6-dimethoxyphenyl-boronic acid,2,6-dimethoxyphenyl boranediol,boronic acid, 2,6-dimethoxyphenyl,pubchem1825,acmc-1clhv,ksc201q0f,2,6-dimethoxy-phenylboronic acid PubChem CID: 2734343 IUPAC Name: (2,6-dimethoxyphenyl)boronic acid SMILES: B(C1=C(C=CC=C1OC)OC)(O)O