accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2305-32-0

HOOOHO

CAS RN 2305-32-0

IUPAC Name: cyclohexane-1,2-dicarboxylic acid
Synonyms: cyclohexane-1,2-dicarboxylic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: QSAWQNUELGIYBC-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results