accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COClSOOCl

CAS RN 22952-32-5

IUPAC Name: 5-chloro-2-methoxybenzene-1-sulfonyl chloride
Synonyms: 5-chloro-2-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 241.08
Molecular Formula: C7H6Cl2O3S
InChi Key: FCJGLIMDVOTBLO-UHFFFAOYSA-N
SMILES: COC1=C(C=C(Cl)C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results