accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOS

CAS RN 22913-24-2

IUPAC Name: methyl 1-benzothiophene-2-carboxylate
Synonyms: methyl 1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 192.23
Molecular Formula: C10H8O2S
InChi Key: KRRAZMUPVIGDCU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results