accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiH3CCH3NCS

CAS RN 2290-65-5

IUPAC Name: isothiocyanatotrimethylsilane
Synonyms: isothiocyanatotrimethylsilane
Molecular Weight (g/mol): 131.27
Molecular Formula: C4H9NSSi
InChi Key: XLTUPERVRFLGLJ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results