accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OSHN

CAS RN 22876-22-8

IUPAC Name: 5-methyl-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 5-methyl-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: SSWZUOXLFTXIEZ-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results