accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 22831-39-6

MgSiMg

CAS RN 22831-39-6

IUPAC Name: dimagnesiosilane
Synonyms: magnesium silicide
Molecular Weight (g/mol): 76.7
Molecular Formula: Mg2Si
InChi Key: YTHCQFKNFVSQBC-UHFFFAOYSA-N
SMILES: [Mg]=[Si]=[Mg]
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results