accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 228244-04-0

CH3H3CCH3OONN

CAS RN 228244-04-0

IUPAC Name: tert-butyl 2-cyanopyrrolidine-1-carboxylate
Synonyms: tert-butyl 2-cyanopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 196.25
Molecular Formula: C10H16N2O2
InChi Key: MDMSZBHMBCNYNO-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results