accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiCH3H3CHNOSiCH3CH3H3C

CAS RN 22737-37-7

IUPAC Name: 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane
Synonyms: 2,2,5,5-tetramethyl-3-oxa-4-aza-2,5-disilahexane
Molecular Weight (g/mol): 177.39
Molecular Formula: C6H19NOSi2
InChi Key: ZAEUMMRLGAMWKE-UHFFFAOYSA-N
SMILES: C[Si](C)(C)NO[Si](C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results