accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHBOHO

CAS RN 227305-69-3

IUPAC Name: (2,3-dihydro-1-benzofuran-5-yl)boronic acid
Synonyms: 2,3-dihydro-1-benzofuran-5-ylboronic acid
Molecular Weight (g/mol): 163.97
Molecular Formula: C8H9BO3
InChi Key: ZIXLJHSFAMVHPC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2OCCC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)