accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 22600-77-7

H2NNHNHClHCl

CAS RN 22600-77-7

IUPAC Name: dihydrogen 1-(1H-imidazol-2-yl)methanamine dichloride
Synonyms: dihydrogen 1-(1H-imidazol-2-yl)methanamine dichloride
Molecular Weight (g/mol): 170.04
Molecular Formula: C4H9Cl2N3
InChi Key: KYUDBQDDNKPSIC-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCC1=NC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results