accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 22600-30-2

CH3OOOH2N

CAS RN 22600-30-2

IUPAC Name: methyl 5-aminofuran-2-carboxylate
Synonyms: methyl 5-aminofuran-2-carboxylate
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: KLKQGSISBRVCTK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results