accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CNCS

CAS RN 2253-73-8

IUPAC Name: 2-isothiocyanatopropane
Synonyms: 2-isothiocyanatopropane
Molecular Weight (g/mol): 101.17
Molecular Formula: C4H7NS
InChi Key: VHBFEIBMZHEWSX-UHFFFAOYSA-N
SMILES: CC(C)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results