CAS RN 22515-18-0
IUPAC Name:
4,4-difluorocyclohexan-1-one
Synonyms:
4,4-difluorocyclohexan-1-one
Molecular Weight (g/mol):
134.13
Molecular Formula:
C6H8F2O
InChi Key:
NYYSPVRERVXMLJ-UHFFFAOYSA-N
SMILES:
FC1(F)CCC(=O)CC1
1
–
2
of
2
results
1
4,4-Difluorocyclohexanone, 97%, Thermo Scientific Chemicals
CAS: 22515-18-0 Molecular Formula: C6H8F2O Molecular Weight (g/mol): 134.126 MDL Number: MFCD08146635 InChI Key: NYYSPVRERVXMLJ-UHFFFAOYSA-N Synonym: 4,4-difluorocyclohexanone,4,4-difluoro-cyclohexanone,cyclohexanone, 4,4-difluoro,difluorocyclohexanone,acmc-1cnv9,4,4-difluoro cyclohexanone,4,4-difluoro-1-cyclohexanone,4,4-difluorocycloxanone,1,1,-difluoro-4-oxocyclohexane PubChem CID: 15789092 IUPAC Name: 4,4-difluorocyclohexan-1-one SMILES: C1CC(CCC1=O)(F)F