accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 223911-98-6

HoBrBrBr

CAS RN 223911-98-6

IUPAC Name: holmium(3+) hydrate tribromide
Synonyms: holmium(3+) hydrate tribromide
Molecular Weight (g/mol): 422.66
Molecular Formula: Br3H2HoO
InChi Key: ZSFPAAHZCBVDRA-UHFFFAOYSA-K
SMILES: O.[Br-].[Br-].[Br-].[Ho+3]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results