accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNCH3OCH3O

CAS RN 2235-46-3

IUPAC Name: N,N-diethyl-3-oxobutanamide
Synonyms: acetoacetamide, N,N-diethyl-
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: NTMXFHGYWJIAAE-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)CC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results