accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2234-14-2

H3CCH3CH3CH3CH3OH3CCH3

CAS RN 2234-14-2

IUPAC Name: 1-[2,4,6-tris(propan-2-yl)phenyl]ethan-1-one
Synonyms: 1-(2,4,6-triisopropylphenyl)ethanone
Molecular Weight (g/mol): 246.39
Molecular Formula: C17H26O
InChi Key: SGRDDUKVLKWIBZ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(C)=O)C(=C1)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results