CAS RN 22236-10-8
CAS RN 22236-10-8
IUPAC Name:
4-(difluoromethoxy)aniline
Synonyms:
4-(difluoromethoxy)aniline
Molecular Weight (g/mol):
159.14
Molecular Formula:
C7H7F2NO
InChi Key:
NDEZTSHWEPQVBX-UHFFFAOYSA-N
SMILES:
NC1=CC=C(OC(F)F)C=C1
1
–
3
of
3
results
3
4-(Difluoromethoxy)aniline, 98%, Thermo Scientific Chemicals
CAS: 22236-10-8 Molecular Formula: C7H7F2NO Molecular Weight (g/mol): 159.13 MDL Number: MFCD00085005 InChI Key: NDEZTSHWEPQVBX-UHFFFAOYSA-N Synonym: 4-difluoromethoxy aniline,4-difluoromethoxy-phenylamine,p-difluoromethoxyaniline,4-amino-alpha,alpha-difluoroanisole,4-difluoromethoxy benzenamine,benzenamine, 4-difluoromethoxy,4-difluoromethoxy phenylamine,4-difluormethoxy anilin,pubchem8467 PubChem CID: 737363 IUPAC Name: 4-(difluoromethoxy)aniline SMILES: C1=CC(=CC=C1N)OC(F)F