accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONHON

CAS RN 2221-13-8

IUPAC Name: 3-phenylimidazolidine-2,4-dione
Synonyms: N-phenyl-2,5-imidazolidione
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: RUEGAVIENIPHGK-UHFFFAOYSA-N
SMILES: O=C1CNC(=O)N1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results