accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2216-92-4

H3COOHN

CAS RN 2216-92-4

IUPAC Name: ethyl 2-(phenylamino)acetate
Synonyms: ethyl 2-(phenylamino)acetate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: MLSGRWDEDYJNER-UHFFFAOYSA-N
SMILES: CCOC(=O)CNC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)