accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOOOHO

CAS RN 2215-89-6

IUPAC Name: 4-(4-carboxyphenoxy)benzoic acid
Synonyms: 4-(4-carboxyphenoxy)benzoic acid
Molecular Weight (g/mol): 258.23
Molecular Formula: C14H10O5
InChi Key: WVDRSXGPQWNUBN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CC=C(C=C2)C(O)=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results