accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClNNCl

CAS RN 2213-63-0

IUPAC Name: 2,3-dichloroquinoxaline
Synonyms: quinoxaline, 2,3-dichloro-
Molecular Weight (g/mol): 199.03
Molecular Formula: C8H4Cl2N2
InChi Key: SPSSDDOTEZKOOV-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)N=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)