CAS RN 22098-10-8
CAS RN 22098-10-8
IUPAC Name:
ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate
Synonyms:
ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate
Molecular Weight (g/mol):
263.11
Molecular Formula:
C8H7BrO3S
InChi Key:
PMBGHMBDWGNJJE-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=O)C1=CC=C(Br)S1
1
–
1
of
1
results
1
ethyle2-(5-bromo-2-thienyl)-2-oxoacetate, 97%, Thermo Scientific™
CAS: 22098-10-8 Molecular Formula: C8H7BrO3S Molecular Weight (g/mol): 263.105 MDL Number: MFCD00085056 InChI Key: PMBGHMBDWGNJJE-UHFFFAOYSA-N Synonym: ethyl 2-5-bromothiophen-2-yl-2-oxoacetate,ethyl 5-bromothien-2-yl glyoxylate,ethyl 2-5-bromo-2-thienyl-2-oxoacetate,ethyl 5-bromothiophene-2-glyoxylate,ethyl 5-bromo-2-thienyl glyoxylate,ethyl 2-5-bromothien-2-yl glyoxalate,ethyl 5-bromothiophen-2-yl oxo acetate,2-bromo-5-ethoxy oxo acetyl thiophene,5-bromothiophen-2-yl oxoacetic acid ethyl ester PubChem CID: 2736376 IUPAC Name: ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate SMILES: CCOC(=O)C(=O)C1=CC=C(S1)Br