accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3FO

CAS RN 22062-53-9

IUPAC Name: 5-fluoro-2-methylbenzaldehyde
Synonyms: 5-fluoro-2-methylbenzaldehyde
Molecular Weight (g/mol): 138.14
Molecular Formula: C8H7FO
InChi Key: MBOXPKNOGZJXPK-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results