accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNHNN(E/Z)

CAS RN 220429-71-0

IUPAC Name: 5-(2-phenylethenyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(2-phenylethenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 172.19
Molecular Formula: C9H8N4
InChi Key: UHYGCMNTYAEAHI-UHFFFAOYSA-N
SMILES: N1N=NC(C=CC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results