accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 219718-86-2

BCH3FFFK(E/Z)

CAS RN 219718-86-2

IUPAC Name: potassium trifluoro[2-(4-methylphenyl)ethenyl]boranuide
Synonyms: potassium trifluoro[2-(4-methylphenyl)ethenyl]boranuide
Molecular Weight (g/mol): 224.07
Molecular Formula: C9H9BF3K
InChi Key: IHPTVBKDBKCIOO-UHFFFAOYSA-N
SMILES: [K+].CC1=CC=C(C=C[B-](F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results