accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HO(S)(R)OH(S)O(S)(S)HOHOOOOOOHHO

CAS RN 21967-41-9

IUPAC Name: (2S,3S,4S,5R,6S)-6-[(5,6-dihydroxy-4-oxo-2-phenyl-4H-chromen-7-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Synonyms: baicalin baicalein 7-O-glucuronide
Molecular Weight (g/mol): 446.36
Molecular Formula: C21H18O11
InChi Key: IKIIZLYTISPENI-ZFORQUDYSA-N
SMILES: O[C@@H]1[C@@H](O)[C@H](OC2=C(O)C(O)=C3C(=O)C=C(OC3=C2)C2=CC=CC=C2)O[C@@H]([C@H]1O)C(O)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (4)
  • (4)