accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21900-42-5

CH3H3CClO

CAS RN 21900-42-5

IUPAC Name: 2,4-dimethylbenzoyl chloride
Synonyms: 2,4-dimethylbenzoyl chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: JUOGRBULLOEKKQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(Cl)=O)C(C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)