accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClOSCl

CAS RN 21815-91-8

IUPAC Name: 3-chloro-1-benzothiophene-2-carbonyl chloride
Synonyms: 3-chloro-1-benzothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 231.09
Molecular Formula: C9H4Cl2OS
InChi Key: GWKSSMDJEWPKCM-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Cl)C2=CC=CC=C2S1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)