accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21762-75-4

ONHSNOO

CAS RN 21762-75-4

IUPAC Name: 7-nitro-3,4-dihydro-2H-1,4-benzothiazin-3-one
Synonyms: 7-nitro-2,4-dihydro-1,4-benzothiazin-3-one
Molecular Weight (g/mol): 210.21
Molecular Formula: C8H6N2O3S
InChi Key: OOICZQBVVODNDJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(NC(=O)CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results