accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NOO

CAS RN 21744-84-3

IUPAC Name: 4-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 4-methyl-2H-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: DBJMEBUKQVZWMD-UHFFFAOYSA-N
SMILES: CN1C(=O)COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results