accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 216854-23-8

H3CH3CCH3OONHHN

CAS RN 216854-23-8

IUPAC Name: tert-butyl N-(piperidin-3-yl)carbamate
Synonyms: tert-butyl N-(piperidin-3-yl)carbamate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: WUOQXNWMYLFAHT-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCCNC1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)