accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 216485-86-8

BOCH3H3CHOOH

CAS RN 216485-86-8

IUPAC Name: [3-(propan-2-yloxy)phenyl]boronic acid
Synonyms: 3-isopropoxyphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: UKZVUHVNTYDSOP-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC(=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results