accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNH2

CAS RN 21627-58-7

IUPAC Name: 2-(pyrrolidin-1-yl)aniline
Synonyms: 2-(pyrrolidin-1-yl)aniline
Molecular Weight (g/mol): 162.24
Molecular Formula: C10H14N2
InChi Key: BFPUBGCFJMIZDF-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results