accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21615-34-9

H3COClO

CAS RN 21615-34-9

IUPAC Name: 2-methoxybenzoyl chloride
Synonyms: 2-methoxybenzoyl chloride
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: RZNHSEZOLFEFGB-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)