accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOOH

CAS RN 215309-00-5

IUPAC Name: 3-(morpholin-4-yl)benzoic acid
Synonyms: 3-(morpholin-4-yl)benzoic acid
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: VSKFQESEPGOWBS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results