CAS RN 21490-63-1
CAS RN 21490-63-1
IUPAC Name:
(2S,3S)-2,3-dimethyloxirane
Synonyms:
(2S,3S)-2,3-dimethyloxirane
Molecular Weight (g/mol):
72.11
Molecular Formula:
C4H8O
InChi Key:
PQXKWPLDPFFDJP-IMJSIDKUSA-N
SMILES:
C[C@@H]1O[C@H]1C
1
–
1
of
1
results
1
Promotions Available
trans-2,3-Epoxybutane, 97%
trans-2,3-Epoxybutane, 97%
CAS: 21490-63-1 Molecular Formula: C4H8O Molecular Weight (g/mol): 72.107 MDL Number: MFCD00005129 InChI Key: PQXKWPLDPFFDJP-QWWZWVQMSA-N Synonym: trans-2,3-dimethyloxirane,trans-2,3-epoxybutane,2r,3r-2,3-dimethyloxirane,oxirane, 2,3-dimethyl-, trans,trans-2-butene oxide,butane, 2,3-epoxy-, trans,e-2r,3r-epoxybutane,trans-,a-butylene oxide,trans-2r,3r-epoxybutane,2,3-dimethyloxirane, trans PubChem CID: 6432237 IUPAC Name: (2R,3R)-2,3-dimethyloxirane SMILES: CC1C(O1)C