accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 214750-75-1

H2COONH(R)OHONHOO

CAS RN 214750-75-1

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(prop-2-en-1-yloxy)carbonyl]amino}hexanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-{[(prop-2-en-1-yloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 452.51
Molecular Formula: C25H28N2O6
InChi Key: OJBNDXHENJDCBA-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)