CAS RN 2144-69-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Bis[p-(2-hydroxyethoxycarbonyl)benzoyloxy]ethane, TRC
CAS: 2144-69-6 Molecular Formula: C22 H22 O10 Molecular Weight (g/mol): 446.404 Synonym: 1,4-Benzenedicarboxylic acid, 1,2-ethanediyl bis(2-hydroxyethyl) ester (9CI),Ethylene glycol, terephthalate (3:2) (8CI),Terephthalic acid, ester with ethylene glycol (2:3) (8CI),Terephthalic acid, ethylene bis(2-hydroxyethyl) ester (6CI, 7CI),1,2-Bis[ IUPAC Name: 1-O-[2-[4-(2-hydroxyethoxycarbonyl)benzoyl]oxyethyl] 4-O-(2-hydroxyethyl) benzene-1,4-dicarboxylate SMILES: OCCOC(=O)c1ccc(cc1)C(=O)OCCOC(=O)c2ccc(cc2)C(=O)OCCO