accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21423-81-4

CH3ClN

CAS RN 21423-81-4

IUPAC Name: 3-chloro-4-methylbenzonitrile
Synonyms: 3-chloro-4-methylbenzonitrile
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: INEMHABDFCKBID-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Cl)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results