accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 21382-98-9

H3CSN

CAS RN 21382-98-9

IUPAC Name: 4-(methylsulfanyl)benzonitrile
Synonyms: 4-(methylsulfanyl)benzonitrile
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: RPVGEEHGKIFQFO-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)